Accessories for HD Super™ Mobile Worktables, Metro International
Catalog #: CA82002-504
Supplier: Metropolitan Wire
Specifications
- Description:Aluminum HD Split Sleeves
- Cat. no.:CA82002-504
- Supplier No.:9986HZ
Specifications
About this item
Aluminum head split sleeves are used to secure Metro HD Super™ shelves to HD Super stationary or mobile posts.
- Required for over-the-road applications and situations where the temperature is above 48°C
- For use with HD Super™ shelving and 15/8" diameter HD posts
Delivery: Sold in pairs of 4. Order one set for each HD Super™ shelf.
Customers Who Bought This Also Bought