Stirring Shaft Coupling, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Universal swivel coupling for connection to a metal chuck
- Stainless steel stir shaft couplings connect glass or metal stir shafts to the adjustable chuck on an overhead stir motor
- Compression connection is secured to various O.D. shafts via an Allen screw coupling
Pass-thru assemblies contain coupling, ⅜" union, and ⅜×12" extension shaft.
The plastic compression connection, secured via an Allen screw, attaches to various O.D. glass stirring shafts. When used with pass-through assemblies, the coupling allows for easy height adjustment flexibility when combined with pass-through capable overhead stirrer motors.