Filtration Apparatus, 25 mm, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
A new concept for lab filtration apparatus from Ace Glass
- Filter unit has four basic parts that can be completely disassembled for easy cleaning
- Instead of a sealed-in glass fritted support, the discs can be easily removed and cleaned
- Discs are also available in different frit sizes, ranging from extra coarse to extra fine
- Autoclavable
For 47 mm version, see 89083-540. For 75 mm version, contact your VWR representative.
Using the famous Ace-Thred design, Ace was able to thread the filter funnel and filter flask together without the silicone stopper and troublesome clamping device. This design also eliminates leakage and alignment issues.
Delivery: Complete Filtration Apparatus includes one each: 500 mL filter flask (89083-542) w/Ace-Thred top, #7 Ace-Thred, and Ace-Safe connector; nylon adapter w/#25 Ace-Threds (89082-470); 25 mm 70-100 micron fritted disc (89083-448), nylon retaining ring (89084-420), 100 mL graduated funnel with Ace-Thred bottom (89083-986). For 500mL filter flask with [ST] 24/40 outer top joint and hose barb side port, order 89057-370. For nylon adapter with #25 Ace-Thred on one end and [ST] 24/40 inner bottom joint, order 89082-476. For 145–174 micron fritted disc, order 89083-446. For 25–50 micron fritted disc, order 89083-450.
Related Information
Ace Glass Filtration Apparatus.pdf Ace-Thred™ Filtraction Apparatus.pdf
Product Literature
Customers Who Bought This Also Bought