BCR® Nylon Glove Liners, Berkshire
Supplier: Berkshire
Certificates
About this item
These lightweight glove liners are made out of high quality 100% contiuous filament pre-shrunk nylon.
- Liners can be re-laundered and used again
- No seams on the gloves that could unravel and cause contamination
- Composition allows moisture to be absorbed and wicked away from skin during extended wear
- Ambidextrous for easy stock control
- Color coded cuffs to identify sizes
The half-finger versions of these liners are ideal for operators who need to retain maximum dexterity and fingertip sensitivity.
Specifications
- Catalog No:
- 14444-322
- 14444-320
- 14444-318
- 14444-316
- 14444-314
- 14444-312
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
Specifications
Related Information
AT-Cleanroom_Poly_Nylon_Glove_Liners
Datasheet
Focus Safety Volume 1 Issue 1 Safety Essentials Volume 2
Product Literature
Customers Who Bought This Also Bought