Closures for RSA™ MRQ™ 1.2 ml Clear Snap/Crimp Top Vial, MicroSolv
Supplier: MicroSolv Technology Corp
Certificates
Closures for RSA™ MRQ™ 1.2 ml Clear Snap/Crimp Top Vial, MicroSolv
Supplier: MicroSolv Technology Corp
About this item
Customers Who Bought This Also Bought
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
$150.16 to $1,502.77
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
n-Pentane ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$761.64 to $2,434.01
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Heavy Duty Aluminum Foil, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$177.29