About this item
The NxGen® T4 DNA Ligases from LGC Biosearch (Lucigen) come in a range of highly pure grades specifically for next-generation sequencing (NGS). This enzyme catalyses the formation of a phosphodiester bond between the terminal 5′ phosphate and 3′ hydroxyl groups of duplex DNA or RNA and efficiently joins blunt and cohesive ends, and repairs single stranded nicks in duplex DNA, RNA, or DNA/RNA hybrids.
- Low concentration ligase 2 U/µL
- High Concentration ligase Rapid Kit 10 U/µL
- NxGen Grade T4 DNA Ligase is ≥ 95% pure
10X T4 DNA Ligase Buffer is provided with T4 DNA Low concentration ligase.
2X Rapid Ligation Buffer and 10X T4 DNA Ligase Buffer is provided with T4 DNA High concentration ligase.
10X T4 DNA Ligase Buffer is composed of 500mM Tris-HCI, 100 mM MgCl2, 50 mM dithiothreitol, 10 mM ATP, pH 7.6 @ 25°C.
2X Rapid Ligation Buffer is composed of 132mM Tris-HCI, 20 mM MgCl2, 2 mM dithiothreitol, 2 mM ATP, 15% PEG, pH 7.6 @ 25°C.
Performance Specifications: 1) SS Exonuclease: 6,000 U <0.1% released. 2) DS Exonuclease: 6,000 U <0.1% released. 3) Endonuclease: 6,000 U <0.1% converted. 4) E. coli 16S rDNA Contamination: 3,000 U < 10 copies. 5) Storage: -20°C.
Source: A recombinant E. coli strain carrying the cloned T4 DNA Ligase gene.