Butyl Rubber Injection Stopper, MicroSolv
Catalog #: 10014-072
Supplier: MicroSolv Technology Corp
Specifications
- Ø:20 mm
- Color:Grey
- Description:Septa, headspace, stopper, for use in 20 mm crimp caps
- Material:Molded butyl rubber
- Cat. no.:10014-072
- Supplier No.:95025-06
Specifications
About this item
Shaped, injection stoppers, butyl rubber used in PerkinElmer Injectors. Use in aluminum or steel crimp caps for headspace autosamplers which are sold separately.
Packaging: Supplied in polyethylene bags.
Customers Who Bought This Also Bought