Methylene blue modified (Newman-Lampert) milk smear stain, for microscopic bacterial examination of milk
Catalog #: RC489016
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number: 61-73-4
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:500 ml
- Cat. no.:RC489016
- Supplier No.:R4890000-500C
Specifications
Specification Test Results
- Appearance:Clear, dark blue, pungent Chloroform-like odor
- Methylene blue modified (Newman-Lampert) milk smear stain, for microscopic bacterial examination of milk:
Specification Test Results
About this item
Synonyms: Basic Blue 9,Methylthioninium chloride,Methylene blue chloride,3,7-Bis(dimethylamino)phenothiazin-5-ium chloride
Boiling Pt:~ 120 °C
Density:1.1620 g/cm³
ADR:3 (6.1),II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: ~ 120 °C
:Melting Pt:
:Melting Pt: 180 °C
:Flash Pt:
:Flash Pt:
:M.W. 319.86 g/mol
:Methylene blue
:180
:~ 120 °C
UN:1992
:3 (6.1)
:II
:GHS08
:GHS06
:GHS05
:GHS09
:GHS02
Formula:C₁₆H₁₈ClN₃S
MW:319.86 g/mol
Storage Temperature:Ambient
:12,06137
MDL Number:MFCD00150008
CAS Number:61-73-4
Customers Who Bought This Also Bought