Brilliant green 1% (w/v) in aqueous solution
Catalog #: RC1155-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number: 633-03-4
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle
- Size:500 ml
- Cat. no.:RC1155-16
- Supplier No.:R1155000-500A
Specifications
Specification Test Results
- Appearance:Clear, dark teal, odorless
- Suitability for Use:Yellow (pH 2.6)
- in aqueous solution:
Specification Test Results
About this item
Synonyms: N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulfate,Ethyl Green,CI 42040,Basic Green 1,N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate,Brilliant Green hydrogen sulfate,Diamond Green,Malachite Green G,Emerald Green,Astradiamant green GX,Solid Green JO
Formula:C₂₇H₃₄N₂O₄S
Boiling Pt:100 °C
Melting Pt:0.0 °C
Density:1.0100 g/cm³
MDL Number:MFCD00011880
CAS Number:633-03-4
:Length
:Width
:Height
:Diameter
:Thickness
:Boiling Pt:
:Boiling Pt: 100 °C
:Melting Pt:
:Melting Pt: 0.0 °C
:Flash Pt:
:Flash Pt:
:M.W. 482.65 g/mol
:100 °C
:GHS07
MW:482.65 g/mol
Storage Temperature:Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
Customers Who Bought This Also Bought