Quick Connect Coupling Assembly for Omni Mixer Homogenizers, Omni International, Inc.
Catalog #: CA11010-582
Supplier: Omni International
Specifications
- Description:Quick Connect Kit (includes Quick Connect Assembly and 3 Quick Connect Adapters).
- Cat. no.:CA11010-582
- Supplier No.:17215
Specifications
About this item
The Quick Connect Assembly (QCA) makes your Omni Mixer homogenizers even more convenient to use
The QCA is easy to install on your existing unit. It comes standard on the Omni Macro-ES. To use, attach a QCA adapter to your chamber assembly or generator probe. Next, slide the adapter in the QCA Slider assembly and a retaining ring positions into place for a secure hold. To disconnect, merely slide the ring upward and the processing accessory is conveniently released.
Ordering information: One quick connect coupling is required per motor unit, and one quick connect adaptor is required per generator probe or assembly.
Customers Who Bought This Also Bought