Calcium chloride dihydrate 97.0-103.0% USP, Ph. Eur., BP, JP, FCC
Catalog #: 89234-902
Supplier: Macco Organiques S.R.O
CAS Number: 10035-04-8
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly drum
- Size:12 kg
- Cat. no.:89234-902
- Supplier No.:52003020120
Specifications
Specification Test Results
- Identification (PhEur/BP/USP/FCC/JP):Corresponds all required tests
- Appearance (PhEur/BP):White or almost shite, crystalline powder.
- Solubility (PhEur/BP):Freely soluble in water, soluble in ethanol (96%)
- Assay (PhEur/BP/USP):97.0 - 103.0%
- Solution clarity (PhEur/BP/JP):Clear
- Solution color (PhEur/BP/JP):Colorless
- Acidity or alkalinity (PhEur/BP):Corresponds to test
- Sulfates (PhEur/BP):0.0003
- Aluminum (PhEur/BP/USP):0.0001% Max.
- Barium (PhEur/BP):Corresponds to test
- Iron (PhEur/BP):0.001% Max.
- Magnesium alkali metals (PhEur/BP):0.5% Max.
- Heavy metals (PhEur/BP):0.002% Max.
- Assay (USP/FCC):99.0-107.0%
- Magnesium alkali metals (USP):1.0% Max.
- Heavy Metals (USP/JP):0.001% Max
- pH (USP/JP):4.5-9.2
- Iron aluminum and phosphate(USP/JP):corresponds to teast
- Organic volatile imputiries (USP):Corresponds to test
- Appearance (FCC):White, hard fragments, granules, or powder.
- Solubility (FCC):Soluble in water and slightly soluble in alcohol.
- Magnesium alkali metals (FCC):4.0% Max.
- Arsenic (FCC):0.003% Max.
- Fluoride (FCC):0.004% Max.
- Lead (FCC):0.0005% Max.
- Appearance (JP):White odorless granules or masses.
- Solubility (JP):Very soluble in water, soluble in ethanol and practically insoluble in diethyl ether.
- Assay (JP):96.7-103.3 %
- Sulfates (JP):0.024% Max
- Arsenic (JP):0.0002% Max (As2O3)
- Hypochlorite (JP):Corresponds to test
- Bromides (available upon request):0.001% Max.
- Particle size greater than 0.800 mm:2-5% (for information only, non-binding)
- Particle size 0.800-0.355 nm:50-80% (for information only, non-binding)
- Particle size less than 0.250 mm:15-48% (for information only, non-binding)
Specification Test Results
About this item
Synonyms: Calcium Chloride Dihydrate
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:1.85 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97,0-103,0
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 176 °C
:Flash Pt:
:Flash Pt:
:M.W. 147.01 g/mol
Merck Index:13,01660
:Calcium chloride dihydrate
Formula:CaCl₂·2H₂O
MW:147.01 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00149613
CAS Number:10035-04-8
EINECS:233-140-8
:GHS07
Customers Who Bought This Also Bought