Magnesium sulfate heptahydrate 99.0-100.5%, crystals USP, Ph. Eur., JP, BP, FCC (GMP, ISO 9001:2000, Kosher, Halal)
Catalog #: 89234-912
Supplier: Macco Organiques S.R.O
CAS Number: 10034-99-8
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:12 kg
- Supplier No.:51204070120
Specifications
Specification Test Results
- Identity Methods A & B (BP/USP/FCC):Pass test
- Appearance (PhEur/BP):White or almost white, crystalline powder or brilliant, colourless crystals
- Solubility (PhEur/BP):Freely soluble in water, very soluble in boiling water, practically, insoluble in ethanol (96 %)
- Assay (PhEur/BP/USP):99.0-100.5% (MgSO4)
- Solution clarity (PhEur/BP/JP):Clear
- Solution color (PhEur/BP/JP):Colorless
- Acidity or alkalinity (PhEur/BP):Corresponds to test
- Chlorides (PhEur/BP):0.03% Max
- Arsenic (PhEur/BP/JP):0.0002% Max.
- Iron (PhEur/BP/USP):0.002% Max.
- Heavy metals (PhEur/BP/USP/JP):0.001% Max.
- Loss on ingintion (PhEur/BP):48.0-52.0%
- Chlorides (USP/JP):0.014% Max.
- Assay (USP/FCC):40.0-52.0%
- pH (USP):5.0-9.2
- Selenium (USP/FCC):0.003% Max.
- Organic volatile impurities (USP):Corresponds to test
- Appearance (FCC):Colorless crystal or a granular crystalline powder.
- Solubility (FCC):Readily soluble in water, slowly soluble in glycerin, and sparingly soluble in alcohol.
- Assay (FCC):99.5% Min. (after ingition)
- Lead (FCC):0.0004% Max.
- Appearance (JP):Colorless or white crystals.
- Solubility (JP):Very soluble in water, and practically insoluble in ethanol 95%.
- Assay (JP):99.0% Min. (MgSO4)
- pH (JP):4.5-8.2
- Loss on ignition (JP):45.0-52.0%
- Zinc (JP):Corresponds to test
- Calcium (JP):Corresponds to test
Specification Test Results
About this item
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 1124 °C
:Flash Pt:
:Flash Pt:
:M.W. 246.48 g/mol
Merck Index:13,05715
Formula:MgSO₄·7H₂O
MW:246.48 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00149785
CAS Number:10034-99-8
EINECS:231-298-2
Customers Who Bought This Also Bought