Zinc sulfate heptahydrate USP, Ph. Eur., FCC
Catalog #: 89234-946
Supplier: Macco Organiques S.R.O
CAS Number: 7446-20-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly drum
- Size:12 kg
- Cat. no.:89234-946
- Supplier No.:53004070120
Specifications
Specification Test Results
- Identity testing (PhEur/USP/FCC):Corresponds to tests
- undefined:White or almost white, crystalline powder, or colorless, transparent crystals.
- Solubility (PhEur):Very soluble in water, practically insoluble in ehtanol 96%
- Assay (PhEur):99.0-104.0%
- Appearance of solution (PhEur):Clear, colorless
- pH (PhEur):4.0-5.7
- Chlorides (PhEur):301 ppm Max.
- Iron (Ph Eur):101 ppm Max.
- Assay (USP):99.0-108.7%
- Alkalies and alkaline earths (USP):0.9% Max.
- Acidity (USP/FCC):Passes test
- Lead (USP):0.002% Max.
- Arsenic (USP):15 ppm Max.
- Appearance (FCC):Colorless, transparent prism, or small needles, or granular crystaline powder.
- Solubility (FCC):Soluble in water and practically insoluble in alcohol.
- Assay (FCC):99.0-108.7%
- Alkalies and alkaline earths (FCC):0.5% Max.
- Lead (FCC):4 mg/kg Max.
- Mercury (FCC):5 mg/kg Max.
- Selenium (FCC):0.003% Max.
- Standard grade particle size greater than 800 micron:10% Max. (information only, non-binding)
- Particle size 355-800 micron:50-70% (information only, non-binding)
- Standard grade particle size less than 355 micron:30-50% (information only, non-binding)
- Fine grade particle size less than 180 micron:99% Min. (information only, non-binding)
Specification Test Results
About this item
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
ADR:9,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 100 °C
:Flash Pt:
:Flash Pt:
:M.W. 287.54 g/mol
Merck Index:12,10293
Formula:ZnSO₄·7H₂O
MW:287.54 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00149894
CAS Number:7446-20-0
EINECS:231-793-3
UN:3077
:9
:III
:GHS05
:GHS07
:GHS09
Customers Who Bought This Also Bought