Specifications
- Description:An Alternative Iodine Clock Reaction
- No. of Lab Groups:15
- Cat. no.:470136-724
- Supplier No.:IS9505
Specifications
About this item
Discover How a Clock Reaction Provides Insight into Reaction Kinetics.
Here’s an easy, effective way to help your students learn the mechanisms and reactions involved in one type of clock reaction – and understand how a clock reaction may provide insight into reaction kinetics. After assembling, performing, and obtaining data from several clock reactions, students will alter experimental conditions and investigate the effects on clock reaction data. Determination of the effects of concentration and temperature on chemical kinetics will also be investigated. Kit contains materials for 15 groups, Teacher’s Manual, Student Study Guide, and copymasters.
- Students use inquiry to determine the effects of environmental conditions
- All chemicals required are included
- Safe alternative to iodine-clock reaction
Here’s an easy, effective way to help your students learn the mechanisms and reactions involved in one type of clock reaction – and understand how a clock reaction may provide insight into reaction kinetics. After assembling, performing, and obtaining data from several clock reactions, students will alter experimental conditions and investigate the effects on clock reaction data. Determination of the effects of concentration and temperature on chemical kinetics will also be investigated.