Specifications
- Description:Green Fuel Cell: Energy From Yeast
- No. of Lab Groups:15
- Cat. no.:470136-738
- Supplier No.:IS9508
Specifications
About this item
Stimulates Discussion About Energy Sources.
The transfer of electrons is part of the metabolic process in living organisms. However, what if it were possible to capture, remove, and use some of these electrons? Could living organisms generate an electrical current? Students will set up a simple cell and, using the easy-to-grow and environmentally tolerant organism yeast, as well as a special dye capable of entering yeast cells and collecting electrons, determine if the harvested electrons are capable of producing current in the cell. If so, how much current? This fun activity also serves as a great tool to stimulate discussion regarding alternate energy sources. Kit contains materials for 15 groups, Teacher’s Manual, Student Study Guide, and copymasters.
- Green chemistry inquiry activity
- All chemicals required are included
- Ideal for biology, biochemistry, earth science and chemistry courses
The transfer of electrons is part of the metabolic process in living organisms. However, what if it were possible to capture, remove, and use some of these electrons? Could living organisms generate an electrical current? Students will set up a simple cell and, using the easy-to-grow and environmentally tolerant organism yeast, as well as a special dye capable of entering yeast cells and collecting electrons, determine if the harvested electrons are capable of producing current in the cell. If so, how much current? This fun activity also serves as a great tool to stimulate discussion regarding alternate energy sources.