Flexam™ Sterile Nitrile Examination Gloves, Cardinal
Supplier: Cardinal Health
Certificates
About this item
Sterile, powder-free nitrile examination gloves are 11.1" in length.
- Ambidexterous
- Size: S to XL
It is textured for a more confident grip.
Packaging: 40 Pairs/box; 5 boxes/case.
Specifications
- Catalog No:
- CAPHN8832
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Disposable