Methanol, anhydrous ≥99.9%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog #: CAMKH08010
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 67-56-1
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, PVC-coated
- Size:4 L
- Cat. no.:CAMKH08010
- Supplier No.:H080-10
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (CH₃OH) (by GC):≥99.9 %
- Carbonyl Compounds - Acetaldehyde:≤0.001 %
- Carbonyl Compounds - Acetone:≤0.001 %
- Carbonyl Compounds - Formaldehyde:≤0.001 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.00010 %
- Solubility in H₂O:Passes Test
- Substances Darkened by H₂SO₄:Passes Test
- Substances Reducing Permanganate:Passes Test
- Titrable Acid (meq/g):≤0.0003
- Titrable Base (meq/g):≤0.0002
- Water (H₂O)(by Karl Fischer titrn):≤0.05 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.80
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.25
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:≤0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:≤0.04
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 - 400 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.005
- Liquid Chromatography Suitability Absorbance:Passes Test
- HPLC Gradient Elution (No Peak Greater than 25% of Full Scale)(0.005 au):Passes Test
- Appearance (clear, colorless liquid):
- Filtered through a 0.2 micron filter.:
- Methanol, anhydrous ≥99.9%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™:
Specification Test Results
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Management of Change (MOC) category = N
Flash Pt:11 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
Merck Index:13,05984
Formula:CH₃OH
MW:32.04 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00004595
CAS Number:67-56-1
EINECS:200-659-6
UN:1230
REACH:01-2119433307-44
:3
:II
:11 °C
:GHS02
:GHS06
:GHS08
:M.O.C. Category N
Customers Who Bought This Also Bought