Dichloromethane ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog #: CAMK488410
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 75-09-2
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth, PVC-coated
- Size:4 L
- Supplier No.:4884-10
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (CH₂Cl₂) (by GC, corrected for water):≥99.5 %
- Appearance:Passes Test
- ACS - Color (APHA):≤10
- ACS - Free Halogens:Passes Test
- ACS - Residue after Evaporation:≤0.002 %
- ACS - Titrable Acid (meq/g):≤0.0003
- ACS - Water (H₂O)(by coulometry):≤0.02 %
- Macron Fine Chemicals™, Dichloromethane ≥99.5% (by GC), AR® ACS:
Specification Test Results
About this item
Synonyms: Methylene chloride
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.32 g/cm³
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = N
Flash Pt:4 °C
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: –95 °C
:Flash Pt:
:Flash Pt: 4 °C
:M.W. 84.93 g/mol
Merck Index:12,09667
:Dichloromethane
Formula:CH₂Cl₂
MW:84.93 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00000881
CAS Number:75-09-2
EINECS:203-625-9
UN:1593
REACH:05-2114615130-69
:6.1
:III
:GHS07
:GHS08
:M.O.C. Category N
Customers Who Bought This Also Bought