Specifications
- Description:Thermal Printer, 115 V
- Cat. no.:CA11500-048
- Supplier No.:89-0069-06
Specifications
About this item
Quickly document gels and blots high quality thermal printer
The thermal printer connects to Analytik Jena BioImaging Systems (such as BioDoc-It® and GelDoc-It™ imagers, except for the PhotoDoc-It™ system) for gel documentation.
Prints are 256-grayscale, publication/archive quality. Two paper types are available for use with printer. The compact print is excellent for limited laboratory bench space. Produces a 75 x 100 mm (3 x 315/16") picture in 1.9 seconds.