Sodium sulfate, anhydrous ≥99.0% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Disodium sulfate,Bisodium sulfate,Disodium sulphate,Sulfuric acid disodium salt,Bisodium sulphate
Formula:Na₂SO₄
MW:142.04 g/mol
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
MDL Number:MFCD00003504
CAS Number:7757-82-6
Merck Index:12,08829
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1429 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 888 °C
:Flash Pt:
:Flash Pt:
:M.W. 142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- Meets E.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (Na₂SO₄) (dried basis):≥99.0 %
- USP - Acidity or Alkalinity:Passes Test
- USP - Chloride (Cl):≤0.005 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - Insoluble Matter:≤0.01 %
- USP - Iron (Fe):≤0.001 %
- USP - Loss on Drying at 105°C:≤0.5 %
- Thru U.S. No. 50 Sieve, For Information Only:
- Solubility (5 in 100):Passes Test
- EP/BP - Assay (Na₂SO₄):98.5 - 101.0 %
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Chloride (Cl):≤450 ppm
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Loss on Drying @ 130° C:≤0.5 %
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- Macron Fine Chemicals™, Sodium sulfate, anhydrous ≥99.0% (dried basis), powder, GenAR® USP for biotechnology:
Specification Test Results
Related Information
Certificates
Search by Lot Number7803 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_MK780388
Product specification
Customers Who Bought This Also Bought