di-Sodium tetraborate decahydrate 99.0-105.0%, GenAR® NF for biotechnology, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Borax tetra
Formula:Na₂B₄O₇·10H₂O
MW:381.37 g/mol
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
MDL Number:MFCD00149193
CAS Number:1303-96-4
Merck Index:13,08662
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-105,0
:Boiling Pt:
:Boiling Pt: 320 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 75 °C
:Flash Pt:
:Flash Pt:
:M.W. 381.37 g/mol
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS07
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets N.F. Requirements:
- Meets E.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- GMP Manufactured Product:
- Preserve in Tight Containers:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (Na₂B₄O₇·10H₂O):99.0 - 105.0 %
- NF - Carbonate and Bicarbonate:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- EP/BP - Assay (Na₂B₄O₇·10H₂O):99.0 - 103.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH of 4% Solution at 25°C:9.0 - 9.6
- EP/BP - Sulfate (SO₄):≤50 ppm
- EP/BP - Ammonium (NH₄):≤10 ppm
- EP/BP - Arsenic (As):≤5 ppm
- EP/BP - Calcium (Ca):≤100 ppm
- JP - Assay (Na₂B₄O₇·10H₂O):99.0 - 103.0 %
- JP - Arsenic (As):≤5 ppm
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - pH (1 in 20):9.1 - 9.6
- JP - Clarity and Color of Solution:Passes Test
- JP - Carbonate and Bicarbonate:Passes Test
- JP - Heavy Metals (as Pb):≤20 ppm
- Appearance (colorless, efflorescent, crystals or crystalline masses or:
- white crystalline powder):
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number7792 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR7792
SDS
spec_MK779288
Product specification
Customers Who Bought This Also Bought