di-Sodium tetraborate decahydrate 99.0-105.0% NF, Macron Fine Chemicals™
Catalog #: CAMK741826
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 1303-96-4
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Fibre drum
- Size:300 lbs.
- Cat. no.:CAMK741826
Specifications
Specification Test Results
- Meets N.F. Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (Na₂B₄O₇·10H₂O):99.0 - 105.0 %
- NF - Carbonate and Bicarbonate:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- Appearance (colorless crystals or white crystalline powder):
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
About this item
Synonyms: Borax tetra
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-105,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 320 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 75 °C
:Flash Pt:
:Flash Pt:
:M.W. 381.37 g/mol
Merck Index:13,08662
Formula:Na₂B₄O₇·10H₂O
MW:381.37 g/mol
MDL Number:MFCD00149193
CAS Number:1303-96-4
EINECS:215-540-4
REACH:01-2119490790-32
:GHS07
:GHS08
Storage Temperature:Ambient
:M.O.C. Category R
Customers Who Bought This Also Bought