Magnesium sulfate heptahydrate 99.0-100.5%, GenAR® USP for parenteral use, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Formula:MgSO₄·7H₂O
MW:246.48 g/mol
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
MDL Number:MFCD00149785
CAS Number:10034-99-8
Merck Index:13,05715
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 1124 °C
:Flash Pt:
:Flash Pt:
:M.W. 246.48 g/mol
Storage Temperature:Ambient
EINECS:231-298-2
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (as MgSO₄):99.0 - 100.5 %
- Appearance:Passes Test
- Solubility (5 in 50):Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH:5.0 - 9.2
- USP - Loss on Ignition:40.0 - 52.0 %
- USP - Chloride (Cl):≤ 0.014 %
- USP - Selenium (Se):≤ 0.003 %
- USP - Iron (Fe):≤ 0.5 ppm
- Assay (MgSO₄):99.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Chloride (Cl):≤ 300 ppm
- EP - Iron (Fe):≤ 20 ppm
- EP/BP - Loss on Drying:48.0 - 52.0 %
- Elemental Impurities (USP (232), EP 5.20) - Information on:
- elemental impurities for this product is available on the:
- associated Product Regulatory Data Sheet and elemental impurity:
- profile report.:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
Specification Test Results
Related Information
Certificates
Search by Lot Number5691 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR5691
SDS
spec_MK569124
Product specification
Customers Who Bought This Also Bought