Calcium chloride, anhydrous 93.0-100.5%, CALTAC FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Chloro calcium
Formula:CaCl₂
MW:110.98 g/mol
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.15 g/cm³ (20 °C)
MDL Number:MFCD00010903
CAS Number:10043-52-4
Merck Index:13,01660
:Length:
:Width:
:Height:
:Ø
:Thickness:
:93,0-100,5
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 772 °C
:Flash Pt:
:Flash Pt:
:M.W. 110.98 g/mol
:Calcium chloride
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
:M.O.C. Category RL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Food GMP Manufactured Product:
- Meets F.C.C. Requirements:
- Preserve in Tight Containers:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- FCC - Assay (CaCl₂):93.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- Solubility:Passes Test
- Acid-Insoluble Substances - Retained Mass:≤0.02 %
- Acid-Insoluble Substances - No Particles >2mm:Passes Test
- Arsenic (As):≤3 mg/kg
- Fluoride (F):≤0.004 %
- FCC - Lead (Pb):≤5 mg/kg
- Magnesium and Alkali Salts:≤5.0 %
- pH (1 in 20):4.5 - 11.0
Specification Test Results
Related Information
Certificates
Search by Lot Number4880 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR4880
SDS
spec_JT4880-32 spec_MK488025
Product specification
Customers Who Bought This Also Bought