Barium sulfate 97.5-100.5% USP suitable for use as excipient, Macron Fine Chemicals™
Catalog #: CAMK451826
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7727-43-7
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Fiber drum
- Size:250 lbs.
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (BaSO₄):97.5 - 100.5 %
- USP - Acid-Soluble Substances:≤0.3 %
- USP - Bulk (fl. oz/lb):20 - 31
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH (10% Suspension):3.5 - 10.0
- USP - Soluble Barium Salts:≤0.001 %
- USP - Sulfide (S):≤0.5 ppm
- Suspension Test for Dirt:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
Specification Test Results
About this item
:Barium sulfate
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:1345 °C
Density:4.5 g/cm³ (25 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97,5-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 1345 °C
:Flash Pt:
:Flash Pt:
:M.W. 233.39 g/mol
Merck Index:13,00997
Formula:BaSO₄
MW:233.39 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00003455
CAS Number:7727-43-7
EINECS:231-784-4
:M.O.C. Category R
Customers Who Bought This Also Bought