Hydrochloric acid 36.5 - 38%, GenAR® NF, Macron Fine Chemicals™
Catalog #: CAMK251546
Supplier: VWR International
CAS Number: 7647-01-0
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear, Mall, PVC-coated
- Size:2.5 L
Specifications
Specification Test Results
- Meets J.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets N.F. Requirements:
- GMP Manufactured Product:
- Store below 25°C:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Identification:Passes Test
- NF - Residue on Ignition:≤ 0.008 %
- NF - Bromide or Iodide:Passes Test
- NF - Free Bromine or Chlorine:Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Sulfite (SO₃):Passes Test
- NF - Assay (HCl)(NF):36.5 - 38.0 %
- Identification A (EP/BP):Passes Test
- Identification B (EP/BP):Passes Test
- Identification C (EP/BP):Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Free Chlorine (as Cl):≤ 4 ppm
- EP/BP - Sulfate (SO₄):≤ 20 ppm
- EP/BP - Residue after Evaporation:≤ 0.01 %
- Assay (HCl)(EP/BP):35.0 - 39.0 %
- JP - Assay (HCl)(JP):35.0 - 38.0 %
- JP - Identification:Passes Test
- JP - Residue on ignition, mg/10mL:≤ 1.0
- JP - Bromide or Iodide:Passes Test
- JP - Free Bromine or Chlorine:Passes Test
- JP - Sulfate (SO₄):Passes Test
- JP - Sulfite (SO₃):Passes Test
- JP - Arsenic (As):≤ 1 ppm
- JP - Heavy Metals (as Pb) (JP):≤ 5 ppm
- JP - Mercury (Hg):≤ 0.04 ppm
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
About this item
Synonyms: Muriatic acid
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 110 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –30 °C
:Flash Pt:
:Flash Pt:
:M.W. 36.46 g/mol
:36.46
Formula:HCl
MW:36.46 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00011324
CAS Number:7647-01-0
EINECS:231-595-7
UN:1789
:8
:II
:GHS05
:GHS07
:M.O.C. Category R
Customers Who Bought This Also Bought