Acetic acid 36% NF, Macron Fine Chemicals™
Catalog #: CAMK248844
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 64-19-7
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear
- Size:2.5 L
- Cat. no.:CAMK248844
- Supplier No.:2488-44
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets N.F. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (CH₃COOH):36.0 - 37.0 %
- NF - Chloride (Cl):Passes Test
- NF - Identification:Passes Test
- NF - Nonvolatile Residue:≤0.005 %
- NF - Readily Oxidizable Substances:Passes Test
- NF - Sulfate (SO₄):Passes Test
- Appearance (clear, colorless liquid):
- Preserve in tight containers, and store at room temperature.:
- Residual Solvents: Only the Class 3 Solvents acetone, isopropyl acetate,:
- and propyl acetate are expected to be present. Residual concentration of all Class 3 Solvents combined is <0.1%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- Macron Fine Chemicals™, Acetic acid 36% NF:
Specification Test Results
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.05 g/mol
EINECS:200-580-7
:17
:118
UN:2790
:8
:III
:38.5
:GHS05
Formula:H₃CCOOH
Storage Temperature:Ambient
:M.O.C. Category R
MW:60.05 g/mol
MDL Number:MFCD00036152
CAS Number:64-19-7
Customers Who Bought This Also Bought