(±)-1,2-Propanediol ≥99.5% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Propylene glycol,1,2-Propanediol,(±)-1,2-Propylene glycol,1,2-Dihydroxypropane,(±)-1
Formula:CH₃CH(OH)CH₂OH
MW:76.1 g/mol
Boiling Pt:187.6 °C (1013 hPa)
Melting Pt:–59 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:99 °C
MDL Number:MFCD00064272
CAS Number:57-55-6
Merck Index:12,08040
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 187.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -59 °C|Melting Pt: –59 °C
:Flash Pt:
:Flash Pt: 99 °C
:M.W. 76.1 g/mol
Storage Temperature:Ambient
EINECS:200-338-0
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- Meets J.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Specific Gravity at 25°/25°C:1.035 - 1.037
- USP - Acidity:Passes Test
- USP - Water (H2O)(by Karl Fischer titrn):≤ 0.20 %
- USP - Identification A (IR):Passes Test
- USP - Identification B - Diethylene Glycol:≤ 0.10 %
- USP - Identification B - Ethylene Glycol:≤ 0.10 %
- USP - Identification C:Passes Test
- USP - Residue on Ignition:≤ 0.007 %
- USP - Chloride (Cl):≤ 0.007 %
- USP - Sulfate (SO₄):≤ 0.006 %
- USP - Assay (CH₃CHOHCH₂OH):≥ 99.5 %
- Endotoxin Concentration (EU/g):≤ 2.5
- EP/BP - Relative Density (d²º₂₀):1.035 - 1.040
- EP/BP - Refractive Index at 20°C:1.431 - 1.433
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Identification D:Passes Test
- EP/BP - Appearance:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Oxidizing Substances:Passes Test
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Water (H₂O):≤ 0.2 %
- EP/BP - Ash (sulfated):≤ 0.01 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Specific Gravity at 20°/20°C:1.035 - 1.040
- JP - Acidity:Passes Test
- JP - Chloride (Cl):≤ 0.007 %
- JP - Sulfate (SO₄):≤ 0.002 %
- JP - Heavy Metals (as Pb):≤ 5 ppm
- JP - Arsenic (As):≤ 2 ppm
- JP - Glycerin:Passes Test
- JP - Water (H₂O):≤ 0.5 %
- JP - Residue on Ignition:≤ 0.005 %
- JP - Distilling Range (Lower):184 - 189 °C
- JP - Distilling Range (Upper):184 - 189 °C
- JP - Ethylene Glycol:≤ 0.1 %
- JP - Diethylene Glycol:≤ 0.1 %
- JP - Any Other Impurity:≤ 0.1 %
- JP - Total Impurities:≤ 1.0 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number9403 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR9403
SDS
spec_JT9403-24 spec_JT9403-3 spec_JT9403-7
Product specification
Customers Who Bought This Also Bought