Ethylene glycol ≥99.0% (by GC), CMOS for microelectronic, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Ethane-1,2-diol,1,2-Dihydroxy ethane,1,2-Ethanediol
Formula:HOCH₂CH₂OH
MW:62.07 g/mol
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:116 °C
MDL Number:MFCD00002885
CAS Number:107-21-1
Merck Index:13,03832
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Boiling Pt:
:Boiling Pt: 196…198 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -13 °C|Melting Pt: –13 °C
:Flash Pt:
:Flash Pt: 116 °C
:M.W. 62.07 g/mol
:Ethylene glycol
Storage Temperature:Refrigerator
EINECS:203-473-3
:196…198 °C (1013 hPa)
REACH:01-2119456816-28
:GHS07
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Microelectronic Use:
- Assay (HOCH₂CH₂OH) (by GC):≥99.0 %
- Color (APHA):≤10
- Acidity (µeq/g):≤0.8
- Acidity (as CH₃COOH)(by wt):≤0.01 %
- Residue after Ignition:≤0.005 %
- Water (H₂O)(by Karl Fischer titrn):≤0.2 %
- Chloride (Cl):≤1 ppm
- Phosphate (PO₄):≤2 ppm
- Sulfate (SO₄):≤2 ppm
- Arsenic and Antimony (as As):≤0.100 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Aluminum (Al):≤100.0 ppb
- Trace Impurities - Barium (Ba):≤100.0 ppb
- Trace Impurities - Boron (B):≤50 ppb
- Trace Impurities - Calcium (Ca):≤300 ppb
- Trace Impurities - Chromium (Cr):≤50 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gold (Au):≤50 ppb
- Trace Impurities - Lead (Pb):≤200.0 ppb
- Trace Impurities - Lithium (Li):≤100.0 ppb
- Trace Impurities - Magnesium (Mg):≤100.0 ppb
- Trace Impurities - Manganese (Mn):≤100.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Sodium (Na):≤300 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤100.0 ppb
- Trace Impurities - Zinc (Zn):≤400.0 ppb
- Particle Count - 0.5 µm and greater:≤200 par/ml
- Particle Count - 1.0 µm and greater:≤10 par/ml
Specification Test Results
Related Information
Certificates
Search by Lot Number9346 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Customers Who Bought This Also Bought