Zinc sulfate heptahydrate ≥99%, granular USP, Multi-Compendial, J.T.Baker®
Catalog #: CAJT4383-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7446-20-0
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. no.:CAJT4383-1
- Supplier No.:4383-01
Specifications
Specification Test Results
- Meets U.S.P Requirements:
- Meets E.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity:Passes Test
- USP - Alkalies and Alkaline Earths:≤0.9 %
- USP - Arsenic (As):≤14 ppm
- USP - Lead (Pb):≤0.002 %
- USP - Assay (ZnSO₄·7H₂O):99.0 - 108.7 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:4.4 - 5.6
- EP/BP - Chloride (Cl):≤300 ppm
- EP/BP - Iron (Fe):≤100 ppm
- EP/BP - Assay (ZnSO₄·7H₂O) (dried basis):99.0 - 104.0 %
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
Specification Test Results
About this item
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
ADR:9,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 100 °C
:Flash Pt:
:Flash Pt:
:M.W. 287.54 g/mol
Merck Index:12,10293
Formula:ZnSO₄·7H₂O
MW:287.54 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00149894
CAS Number:7446-20-0
EINECS:231-793-3
UN:3077
:9
:III
:GHS05
:GHS07
:GHS09
:M.O.C. Category R
Customers Who Bought This Also Bought