Zinc acetate dihydrate 98.0-102.0%, crystals, Technical Grade, J.T.Baker®
Catalog #: CAJT4304-R
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 5970-45-6
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Fiber drum
- Size:250 lbs.
- Cat. no.:CAJT4304-R
- Supplier No.:4304-R
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay ((CH₃COO)₂Zn·2H₂O):98.0 - 102.0 %
- Insoluble Matter:≤0.01 %
- pH of 5% Solution at 25°C:6.0 - 7.0
- Lead (Pb):≤0.005 %
Specification Test Results
About this item
Synonyms: Acetic acid zinc salt dihydrate,Zinc (II) acetate dihydrate
Melting Pt:237 °C
Density:1.718 g/cm³ (20 °C)
ADR:9,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-102,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 237 °C
:Flash Pt:
:Flash Pt:
:M.W. 219.51 g/mol
Merck Index:12,10256
Formula:(H₃CCOO)₂Zn·2H₂O
MW:219.51 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00066961
CAS Number:5970-45-6
EINECS:209-170-2
UN:3077
:9
:III
:GHS07
:GHS09
:M.O.C. Category N
Customers Who Bought This Also Bought