Sodium carbonate 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Soda,Carbonic acid sodium salt,Disodium carbonate,Na-X,Bisodium carbonate,Carbonic acid, disodium salt
Formula:Na₂CO₃
MW:105.99 g/mol
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2.53 g/cm³
MDL Number:MFCD00003494
CAS Number:497-19-8
Merck Index:14,08596
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 854 °C
:Flash Pt:
:Flash Pt:
:M.W. 105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets N.F. Requirements:
- Meets J.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Water (H₂O):≤ 0.5 %
- NF - Assay (Na₂CO₃) (anhydrous basis):99.5 - 100.5 %
- EP/BP - Assay (Na₂CO₃) (dried basis):99.5 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Identification D:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Alkali Hydroxides and Bicarbonates:Passes Test
- EP/BP - Chloride (Cl):≤ 125 ppm
- EP/BP - Sulfate (SO₄):≤ 250 ppm
- EP/BP - Iron (Fe):≤ 50 ppm
- EP/BP - Loss on Drying:≤ 1.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.071 %
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Arsenic (As):≤ 3.1 ppm
- JP - Loss on Drying:≤ 0.5 %
- JP - Assay (Na₂CO₃) (dried basis):99.0 - 101.0 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium carbonate 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number3606 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR3606
SDS
spec_JT3606-7 spec_JT3606-9 spec_JT3606-R
Product specification
Customers Who Bought This Also Bought