Potassium acetate ≥99%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Acetic acid potassium salt
Formula:CH₃COOK
MW:98.14 g/mol
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
MDL Number:MFCD00012458
CAS Number:127-08-2
Merck Index:12,07764
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = NI
Flash Pt:>250 °C
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 304 °C
:Flash Pt:
:Flash Pt: >250 °C
:M.W. 98.14 g/mol
EINECS:204-822-2
:>250 °C
Storage Temperature:Ambient
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (CH₃COOK) (by non-aqueous titration):≥99.0 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:6.5 - 9.0
- Chloride (Cl):≤0.003 %
- Phosphate (PO₄):≤0.001 %
- Sulfate (SO₄):≤0.002 %
- Calcium (Ca):≤0.005 %
- ACS - Magnesium (Mg):≤0.002 %
- Sodium (Na):≤0.03 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Potassium acetate ≥99%, crystals, BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number2912 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR2912
SDS
Customers Who Bought This Also Bought