Sodium hydroxide 0.5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Caustic soda
Formula:NaOH
MW:40 g/mol
Boiling Pt:105 °C (1013 hPa)
Density:1.02 g/cm³ (25 °C)
MDL Number:MFCD00003548
CAS Number:1310-73-2
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 105 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 323 °C
:Flash Pt:
:Flash Pt:
:M.W. 40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
:323
:105 °C (1013 hPa)
UN:1824
:8
:II
:GHS05
:M.O.C. Category R
:12,08772
:14,08627
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Protect from air to avoid absorption of carbon dioxide.:
- CAUTION: For Manufacturing, processing or repackaging:
- Appearance (clear, colorless solution):Passes Test
- Normality:0.480 - 0.520
- Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
- Endotoxin Concentration (EU/mL):≤2.0
- Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications:
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- Filtered through a 0.2 micron filter.:
- in aqueous solution:
Specification Test Results
Related Information
Certificates
Search by Lot Number0329 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT0329-26 spec_JT0329-7
Product specification
Customers Who Bought This Also Bought