L(+)-Cysteine hydrochloride monohydrate 98.5-101.0% (alkalimetric, calculated on anhydrous substance), EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate,(R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate,H-Cys-OH.HCl.H2O,Cysteine Hydrochloride, Monohydrate
Formula:HSCH₂CH(NH₂)COOH·HCl·H₂O
MW:175.64 g/mol
Melting Pt:168…170 °C
Density:1.54 g/cm³
MDL Number:MFCD00065606
CAS Number:7048-04-6
Merck Index:13,02810
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-101,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 168 to 170 °C|Melting Pt: 168…170 °C
:Flash Pt:
:Flash Pt:
:M.W. 175.64 g/mol
:L(+)-Cysteine hydrochloride monohydrate
EINECS:200-157-7
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (Iodometric, calc. on Dried Substance):98.5-101.0%
- Ammonium (NH4):Max. 0.02%
- Appearance:White Crystalline Powder or Colorless Crystals
- Appearance of Solution (25 g/l, water):Clear and no more Intense in Color than BY6
- Heavy metals (as Pb):Max 0.001%
- Identity (IR-spectrum):Passes test
- Iron (Fe):Max. 0.002%
- Loss on Drying (Vacuum):Max. 12.0%
- Ninhydrin-Positive Substances (TLC):Max. 0.5%
- Organic Volatile Impurities (According to USP):Conforms
- Residual Solvents (Ph.Eur./ICH):Excluded by Manufacturing Process
- Specific Rotation [a]20/D; 80 g/l; HCl 250 g/l, Dried:+5.5 - +7.0°
- Specific rotation [a]25/D; 80g/l; HCl 6mol/l, Dried:+5.7 - +6.8°
- Sulfate (SO4):Max. 0.03%
- Sulfated ash (600 °C):Max. 0.1%
- Corresponds to Ph. Eur., USP:
Specification Test Results
Customers Who Bought This Also Bought