Staining Dish, 8-16 Slide Unit, Wheaton
Catalog #: 25461-034
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Dimensions:76 L x 25 W x 75 H mm
- Description:Staining Dish, 8-16 Slide Unit
- Supplier No.:900630
Specifications
About this item
Customers Who Bought This Also Bought