To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Animal-free:Y
- Protease-free:Y
- Protein/peptide type:Recombinant
- Source:Serum
- Species:Human
- Size:1 g
- Storage conditions:Refrigerate/Frozen
- Protein synonyms:Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
- Protein/peptide name:Albumin
- Shipping temperature:Ambient
- Cat. no.:CA89133-078
- Supplier No.:62-450-RF
Specifications
About this item
Corning® cellgro® Albumin is a chemically-defined recombinant human serum albumin (rhHSA). Having an essential and non-essential fatty acid profile, this is a powerful media supplement that outperforms plasma-derived human serum albumin (pHSA), bovine serum albumin (BSA) and FBS. Some prominent applications for cellgro® rhAlbumin are as a supplement for CHO cell culture and for the culture of various other cell lines. This product is plant-derived and completely animal-origin free.