Sampling Adapter with Septa, Ace Glass Incorporated
Catalog #: 89064-900
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Sampling Adapter with Serum Top, [ST] 14/20 Inner Bottom, Septum Included
- Cat. no.:89064-900
- Supplier No.:9096-06
Specifications
About this item
This sampling adapter comes complete with a matching septum.
- Bottom ground [ST] inner joint
- Top will accept included septum
- Glassware portion manufactured from borosilicate glass
The bottom of the adapter is a ground [ST] 14/20 inner joint, and the top will accept the included septum. Septum is white in color, and is a sleeve type that folds down over the neck of the adapter. Diaphragm of the septum can be punctured with a syringe needed, and the puncture seals automatically after the needle is withdrawn.
Customers Who Bought This Also Bought