Immunotechnology Activity Kit
Immunotechnology Activity Kit, Immunoprecipitation Technique
Catalog # 470150-030
Supplier: G-Biosciences
Supplier #: BE-506
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Hands-on Study of Immunoprecipitation
This kit contains all the reagents necessary to immunoprecipitate a specific protein from a complex sample. This hands-on activity focusing on immunoprecipitation permits the purification of specific proteins for which an antibody has been raised. The kit includes enough materials for six groups of four students to complete the experiment.
Specifications
- Description:Immunotechnology Activity Kit, Immunoprecipitation Technique