Power Hold Clamp, Ace Glass Incorporated
Catalog #: 89075-632
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Power Hold Clamp
- Cat. no.:89075-632
- Supplier No.:11082-07
Specifications
About this item
This clamp fits a support stand with 3/8" to 5/8" diameter shaft, and stirrers with mounting rod from 3/8" to 5/8" diameter
Stop collar included.
Customers Who Bought This Also Bought