Tubing Clamp, Ace Glass Incorporated
Catalog #: 89075-638
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Description:Tubing Clamp
- Cat. no.:89075-638
- Supplier No.:11145-20
Specifications
About this item
This tubing clamp eliminates the older method of wiring rubber tubing to glass
Simple in design and easy to use. Requires only one hand to apply; can be instantly applied or removed. Fits standard 10–12mm fittings.
Customers Who Bought This Also Bought