Hosecock, Ace Glass Incorporated
Catalog #: 89084-178
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Hosecock, Standard
- Cat. no.:89084-178
- Supplier No.:11140-25
Specifications
About this item
This hosecock is used to control the flow of liquids or gases through flexible tubing
Single adjustment screw for easy operation. Convex bearing surfaces and rounded edges protect tubing. Machined to deliver accurate regulation of fluid flow or interruption of flow.
The large compression screw permits more precise control than ordinary clamps. In the open position, it can be slipped over tubing which is part of an assembly. Accommodates tubing up to 9.5mm O.D.
Customers Who Bought This Also Bought