Midget Bubbler, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
The nozzle on this midget bubbler has a sintered glass filter, Porosity A (145-175 microns)
Used in determination of Acrolein: 4-Hexylresorcinol Method #1. Bottle has encircling graduations from 0 to 25mL or 0 to 30mL in 5mL divisions. Inlet/outlet are 7mm O.D. Fabricated from borosilicate glass.
Ordering information: Available complete with bottle and tube, or separately.
Customers Who Bought This Also Bought