Pluro Stopper Set, Ace Glass Incorporated
Catalog #: 89075-804
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Pluro Stopper Set
- Cat. no.:89075-804
- For O.D.:18-70 mm
- Supplier No.:12014-14
Specifications
About this item
Versatile silicone stopper set that equals 17 standard stoppers
All the rings are cut from the same stopper, with each ring nesting perfectly into the next. Whether making up a small or large stopper, a vacuum-tight fit is assured. Sold as a set.
Customers Who Bought This Also Bought