ProFlex® 230 Wide Soft Cap Knee Pads, Ergodyne®
Catalog #: 56618-274
Supplier: Ergodyne
Specifications
- Color:Black
- Size:Universal
- Cat. no.:56618-274
- Supplier No.:18230
Specifications
About this item
Knee pads provide economical, light-duty protection for landscaping, maintenance, and a wide range of additional applications
They feature thick 15mm closed cell foam padding and a durable 600D polyester fabric cover. Designed slightly wider and longer than standard knee pads for wraparound comfort, they are available in one easily adjustable size for an exact fit. Dry surface technology is incorporated for optimal grip in dry conditions. Buckle closure. Hand wash.
Customers Who Bought This Also Bought