Dithiothreitol (DTT, Cleland's reagent)
Catalog #: IB21040
Supplier: IBI Scientific
CAS Number: 3483-12-3
Specifications
- Size:5 g
- Cat. no.:IB21040
- Supplier No.:21040
Specifications
Specification Test Results
- -SH content Min.:99.4%
- A283 (0.02 M, H2O) Max.:0.05
- -S-S-content Max.:0.5%
- Melting Point:41 ± 2°C
- IR:Pass
- Moisture Max:0.5%
- Solubility (5%, water):Clear and Colorless
Specification Test Results
About this item
Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol,(R*,R*)-1,4-Dimercapto-2,3-butandiol,DTT,DL-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 42 to 44 °C
:Flash Pt:
:Flash Pt:
:M.W. 154.25 g/mol
:Dithiothreitol (DTT, Cleland's reagent)
:GHS07
MW:154.25 g/mol
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
Customers Who Bought This Also Bought