X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Catalog #: 97060-740
Supplier: IBI Scientific
CAS Number: 7240-90-6
Specifications
- Size:25 g
- Cat. no.:97060-740
- Supplier No.:IB02265
Specifications
About this item
Synonyms: BCIG,(2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 237 to 239 °C
:Flash Pt:
:Flash Pt:
:M.W. 408.63 g/mol
:X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula:C₁₄H₁₅BrClNO₆
MW:408.63 g/mol
MDL Number:MFCD00005666
CAS Number:7240-90-6
Customers Who Bought This Also Bought