Axygen® Automation Tips for Qiagen BioRobot® Series, Corning
Supplier: Corning
Certificates
About this item
Axygen® offers a comprehensive line of tips for Qiagen® automation systems.
- Black tips are conductive for liquid level sensing protocols
- Optimised fine point for accurate dispensing
- Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Specifications
- Catalog No:
- 89124-962
- 89124-978
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Related Information
Certificates
Search by Lot Number89124-978_09219039 89124-978_09219040 89124-978_09519056
Conformity declaration
Corning Automation Tip Compatibility Chart
Product Literature
Customers Who Bought This Also Bought