Formaldehyde 10% (v/v) in aqueous solution stabilized tissue fixative, for light and electron microscopy, buffered (Carson-Millonig Form. pH 7.4)
Supplier: Ricca Chemical
Certificates
About this item
Synonyms: Paraformaldehyde aqueous solution,Methanal solution,Methylaldehyde solution,Formol,Formaline solution,Formalin
Formula:CH₂O
MW:30.03 g/mol
MDL Number:MFCD00003274
CAS Number:50-00-0
ADR:9,
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -92 °C
:Flash Pt:
:Flash Pt:
:M.W. 30.03 g/mol
EINECS:200-001-8
:-92
:-19.3
UN:3334
:9
:GHS06
:GHS08
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:Clear, colorless, Formaldehyde odor
- pH at 25 °C (traceable to NIST SRM 186 & 191):7.40 ± 0.05 pH at 25.0 °C
- Formaldehyde 10% (v/v) in aqueous solution stabilized tissue fixative, for light and electron microscopy, buffered (Carson-Millonig Form. pH 7.4):
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R3191000
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought