Butyl Rubber Two-Leg Stopper, Wheaton
Catalog #: 89050-894
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Color:Gray
- Material:Butyl rubber
- Size:11 mm
- Cat. no.:89050-894
- Supplier No.:W651450
Specifications
About this item
Lyophillization stopper made from grey butyl rubber, for use with WHEATON® Vacule® Lyophillization ampule.
- 2-leg style
- Autoclavable
Customers Who Bought This Also Bought